missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 148270050

 View more versions of this product

Catalog No. AC148270050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Dimethylaminopyridine
98.5
110.0°C to 113.0°C
124°C
Glass bottle
(C5H4N)N(CH3)2
5 g
03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
VHYFNPMBLIVWCW-UHFFFAOYSA-N
N,N-dimethylpyridin-4-amine
14284
Negligible
1122-58-3
100.0
162.0°C (50.0 mmHg)
98.5% min. (GC)
C7H10N2
MFCD00006418
22, II, 341
15, 3433
Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform
CN(C)C1=CC=NC=C1
122.17
122.17
99%

Chemical Identifiers

1122-58-3
VHYFNPMBLIVWCW-UHFFFAOYSA-N
14284
CN(C)C1=CC=NC=C1
MFCD00006418
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
N,N-dimethylpyridin-4-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes skin irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 214-353-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only