missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, 99%

CAS: 1122-58-3 | C7H10N2 | 122.17 g/mol

$50.48 - $1297.01

Chemical Identifiers

CAS 1122-58-3
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC148270050
SDS View Documents
Thermo Scientific Chemicals
148270050
5 g Glass bottle
Each for $50.48
Only null left
 
AC148270250
SDS View Documents
Thermo Scientific Chemicals
148270250
25 g Glass bottle
Each for $131.20
Only null left
 
AC148271000
SDS View Documents
Thermo Scientific Chemicals
148271000
100 g Glass Bottle
Each for $370.23
Only null left
 
AC148275000
SDS View Documents
Thermo Scientific Chemicals
148275000
500 g Glass bottle
Each for $1,297.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1122-58-3
VHYFNPMBLIVWCW-UHFFFAOYSA-N
14284
CN(C)C1=CC=NC=C1
MFCD00006418
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
N,N-dimethylpyridin-4-amine

Specifications

1122-58-3
100.0
162.0°C (50.0 mmHg)
98.5% min. (GC)
C7H10N2
MFCD00006418
22, II, 341
15, 3433
Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform
CN(C)C1=CC=NC=C1
122.17
122.17
99%
98.5
110.0°C to 113.0°C
124°C
Glass bottle
(C5H4N)N(CH3)2
5 g
03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
VHYFNPMBLIVWCW-UHFFFAOYSA-N
N,N-dimethylpyridin-4-amine
14284
Negligible
4-Dimethylaminopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes skin irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 214-353-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only