Learn More
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148271000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Dimethylaminopyridine | |
98.5 | |
99% | |
C7H10N2 | |
MFCD00006418 | |
22, II, 341 | |
15, 3433 | |
VHYFNPMBLIVWCW-UHFFFAOYSA-N | |
N,N-dimethylpyridin-4-amine | |
14284 | |
Negligible |
1122-58-3 | |
100.0 | |
Glass Bottle | |
(C5H4N)N(CH3)2 | |
100 g | |
03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142 | |
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine | |
CN(C)C1=CC=NC=C1 | |
122.171 | |
122.17 |
Chemical Identifiers
1122-58-3 | |
VHYFNPMBLIVWCW-UHFFFAOYSA-N | |
14284 | |
CN(C)C1=CC=NC=C1 |
MFCD00006418 | |
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine | |
N,N-dimethylpyridin-4-amine |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only