Learn More
4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148275000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Dimethylaminopyridine | |
98.5 | |
110.0°C to 113.0°C | |
124°C | |
Glass bottle | |
(C5H4N)N(CH3)2 | |
500 g | |
03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142 | |
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine | |
VHYFNPMBLIVWCW-UHFFFAOYSA-N | |
N,N-dimethylpyridin-4-amine | |
14284 | |
Negligible |
1122-58-3 | |
100.0 | |
162.0°C (50.0 mmHg) | |
98.5% min. (GC) | |
C7H10N2 | |
MFCD00006418 | |
22, II, 341 | |
15, 3433 | |
Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform | |
CN(C)C1=CC=NC=C1 | |
122.17 | |
122.17 | |
99% |
Chemical Identifiers
1122-58-3 | |
VHYFNPMBLIVWCW-UHFFFAOYSA-N | |
14284 | |
CN(C)C1=CC=NC=C1 |
MFCD00006418 | |
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine | |
N,N-dimethylpyridin-4-amine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes skin irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 214-353-5
RUO – Research Use Only