Learn More
4-Ethyltoluene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0212022
Description
4-Ethyltoluene is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethyltoluene is used as intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
4-Ethyltoluene | |
-62°C | |
161°C to 163°C | |
C9H12 | |
MFCD00009263 | |
UN3295 | |
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene | |
JRLPEMVDPFPYPJ-UHFFFAOYSA-N | |
1-ethyl-4-methylbenzene | |
12160 | |
97% |
622-96-8 | |
0.861 | |
43°C (109°F) | |
1.495 | |
100 g | |
1903007 | |
Insoluble in water. | |
CCC1=CC=C(C=C1)C | |
120.195 | |
120.2 |
Chemical Identifiers
622-96-8 | |
120.195 | |
JRLPEMVDPFPYPJ-UHFFFAOYSA-N | |
12160 | |
CCC1=CC=C(C=C1)C |
C9H12 | |
MFCD00009263 | |
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene | |
1-ethyl-4-methylbenzene |
Safety and Handling
GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H226-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 210-761-2
RTECSNumber : XT2550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only