Learn More
4-Ethyltoluene, 97%, Thermo Scientific Chemicals
$71.59 - $516.81
Chemical Identifiers
CAS | 622-96-8 |
---|---|
Molecular Formula | C9H12 |
Molecular Weight (g/mol) | 120.195 |
MDL Number | MFCD00009263 |
InChI Key | JRLPEMVDPFPYPJ-UHFFFAOYSA-N |
Synonym | 4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene |
PubChem CID | 12160 |
IUPAC Name | 1-ethyl-4-methylbenzene |
SMILES | CCC1=CC=C(C=C1)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0212006
|
Thermo Scientific Chemicals
L0212006 |
5 g |
N/A
|
|
||||||
AAL0212014
|
Thermo Scientific Chemicals
L0212014 |
25 g |
N/A
|
|
||||||
AAL0212022
|
Thermo Scientific Chemicals
L0212022 |
100 g |
N/A
|
|
||||||
Description
4-Ethyltoluene is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethyltoluene is used as intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Chemical Identifiers
622-96-8 | |
120.195 | |
JRLPEMVDPFPYPJ-UHFFFAOYSA-N | |
12160 | |
CCC1=CC=C(C=C1)C |
C9H12 | |
MFCD00009263 | |
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene | |
1-ethyl-4-methylbenzene |
Specifications
622-96-8 | |
0.861 | |
43°C (109°F) | |
1.495 | |
5 g | |
1903007 | |
Insoluble in water. | |
CCC1=CC=C(C=C1)C | |
120.195 | |
120.2 | |
4-Ethyltoluene |
-62°C | |
161°C to 163°C | |
C9H12 | |
MFCD00009263 | |
UN3295 | |
4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene | |
JRLPEMVDPFPYPJ-UHFFFAOYSA-N | |
1-ethyl-4-methylbenzene | |
12160 | |
97% |
Safety and Handling
GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H226-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 210-761-2
RTECSNumber : XT2550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only