missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-methylaniline, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1548214

 View more versions of this product

Catalog No. AAA1548214


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluoro-3-methylaniline
32°C to 36°C
85°C to 86°C (9 mmHg)
Characteristic
1.537
25 g
2935561
NYMDPDNETOLVBS-UHFFFAOYSA-N
4-fluoro-3-methylaniline
67981
97%
452-69-7
1.12
95°C (203°F)
C7H8FN
MFCD00025294
UN2811
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
CC1=CC(N)=CC=C1F
125.15
125.15

Chemical Identifiers

452-69-7
125.15
NYMDPDNETOLVBS-UHFFFAOYSA-N
67981
CC1=CC(N)=CC=C1F
C7H8FN
MFCD00025294
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
4-fluoro-3-methylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 207-207-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only