missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluoro-3-methylaniline, 97%, Thermo Scientific Chemicals

$111.70 - $339.40

Chemical Identifiers

CAS 452-69-7
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00025294
InChI Key NYMDPDNETOLVBS-UHFFFAOYSA-N
Synonym 2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
PubChem CID 67981
IUPAC Name 4-fluoro-3-methylaniline
SMILES CC1=CC(N)=CC=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1548206
View Documents
Thermo Scientific Chemicals
A1548206
5 g
Each for $111.70
 
AAA1548214
View Documents
Thermo Scientific Chemicals
A1548214
25 g
Each for $339.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

452-69-7
125.15
NYMDPDNETOLVBS-UHFFFAOYSA-N
67981
CC1=CC(N)=CC=C1F
C7H8FN
MFCD00025294
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
4-fluoro-3-methylaniline

Specifications

452-69-7
1.12
95°C (203°F)
C7H8FN
MFCD00025294
UN2811
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
CC1=CC(N)=CC=C1F
125.15
125.15
4-Fluoro-3-methylaniline
32°C to 36°C
85°C to 86°C (9 mmHg)
Characteristic
1.537
5 g
2935561
NYMDPDNETOLVBS-UHFFFAOYSA-N
4-fluoro-3-methylaniline
67981
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 207-207-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only