missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorophenyl Chlorothionoformate 98.0+%, TCI America™

Supplier:  TCI America F03655G

 View more versions of this product

Catalog No. F03655G


Explore available promotions

Description

Description

Specifications

Specifications

4-Fluorophenyl Chlorothionoformate
Yellow
C7H4ClFOS
5 g
Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate
FC1=CC=C(OC(Cl)=S)C=C1
190.62
190.62
Liquid
42908-73-6
82°C
MFCD00134402
1760
HZBXSUFBKXPOAF-UHFFFAOYSA-N
4-fluorophenyl chloromethanethioate
5045332
≥98.0% (T)

Chemical Identifiers

42908-73-6
190.62
HZBXSUFBKXPOAF-UHFFFAOYSA-N
5045332
FC1=CC=C(OC(Cl)=S)C=C1
C7H4ClFOS
MFCD00134402
Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate
4-fluorophenyl chloromethanethioate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions