missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Fluorophenyl Chlorothionoformate 98.0+%, TCI America™
$98.75 - $292.30
Chemical Identifiers
CAS | 42908-73-6 |
---|---|
Molecular Formula | C7H4ClFOS |
Molecular Weight (g/mol) | 190.62 |
MDL Number | MFCD00134402 |
InChI Key | HZBXSUFBKXPOAF-UHFFFAOYSA-N |
Synonym | Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate |
PubChem CID | 5045332 |
IUPAC Name | 4-fluorophenyl chloromethanethioate |
SMILES | FC1=CC=C(OC(Cl)=S)C=C1 |
Chemical Identifiers
42908-73-6 | |
190.62 | |
HZBXSUFBKXPOAF-UHFFFAOYSA-N | |
5045332 | |
FC1=CC=C(OC(Cl)=S)C=C1 |
C7H4ClFOS | |
MFCD00134402 | |
Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate | |
4-fluorophenyl chloromethanethioate |
Specifications
42908-73-6 | |
82°C | |
MFCD00134402 | |
1760 | |
HZBXSUFBKXPOAF-UHFFFAOYSA-N | |
4-fluorophenyl chloromethanethioate | |
5045332 | |
≥98.0% (T) | |
4-Fluorophenyl Chlorothionoformate |
Yellow | |
C7H4ClFOS | |
1 g | |
Chlorothioformic Acid O-(4-Fluorophenyl) Ester, O-(4-Fluorophenyl) Chlorothioformate | |
FC1=CC=C(OC(Cl)=S)C=C1 | |
190.62 | |
190.62 | |
Liquid |
Safety and Handling
TSCA : No