missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3209303

 View more versions of this product

Catalog No. AAH3209303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Fluorothiobenzamide
146°C to 148°C
MFCD01313285
UN2811
VQFOHZWOKJQOGO-UHFFFAOYSA-N
4-fluorobenzenecarbothioamide
737223
97%
22179-72-2
C7H6FNS
1 g
4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide
NC(=S)C1=CC=C(F)C=C1
155.19
155.19

Chemical Identifiers

22179-72-2
155.19
VQFOHZWOKJQOGO-UHFFFAOYSA-N
737223
NC(=S)C1=CC=C(F)C=C1
C7H6FNS
MFCD01313285
4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide
4-fluorobenzenecarbothioamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only