Learn More
4-Fluorothiobenzamide, 97%, Thermo Scientific Chemicals
$64.73 - $218.12
Chemical Identifiers
CAS | 22179-72-2 |
---|---|
Molecular Formula | C7H6FNS |
Molecular Weight (g/mol) | 155.19 |
MDL Number | MFCD01313285 |
InChI Key | VQFOHZWOKJQOGO-UHFFFAOYSA-N |
Synonym | 4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide |
PubChem CID | 737223 |
IUPAC Name | 4-fluorobenzenecarbothioamide |
SMILES | NC(=S)C1=CC=C(F)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3209303
|
Thermo Scientific Chemicals
H3209303 |
1 g |
Each for $64.73
|
|
|||||
AAH3209306
|
Thermo Scientific Chemicals
H3209306 |
5 g |
Each for $218.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22179-72-2 | |
155.19 | |
VQFOHZWOKJQOGO-UHFFFAOYSA-N | |
737223 | |
NC(=S)C1=CC=C(F)C=C1 |
C7H6FNS | |
MFCD01313285 | |
4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide | |
4-fluorobenzenecarbothioamide |
Specifications
22179-72-2 | |
C7H6FNS | |
1 g | |
4-fluorothiobenzamide, 4-fluorobenzothioamide, 4-fluorobenzene-1-carbothioamide, 4-fluoro-1-benzenecarbothioamide, 4-fluoro-thiobenzamide, benzenecarbothioamide, 4-fluoro, amino 4-fluorophenyl methane-1-thione, p-fluorothiobenzamide, acmc-20amhq, 4-fluoro thiobenzamide | |
NC(=S)C1=CC=C(F)C=C1 | |
155.19 | |
155.19 | |
4-Fluorothiobenzamide |
146°C to 148°C | |
MFCD01313285 | |
UN2811 | |
VQFOHZWOKJQOGO-UHFFFAOYSA-N | |
4-fluorobenzenecarbothioamide | |
737223 | |
97% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only