missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Formylphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 87199-17-5
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151823
InChI Key VXWBQOJISHAKKM-UHFFFAOYSA-N
Synonym 4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
PubChem CID 591073
IUPAC Name (4-formylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359380010
SDS View Documents
Thermo Scientific Chemicals
359380010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359380050
SDS View Documents
Thermo Scientific Chemicals
359380050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359380250
SDS View Documents
Thermo Scientific Chemicals
359380250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

87199-17-5
149.94
VXWBQOJISHAKKM-UHFFFAOYSA-N
591073
OB(O)C1=CC=C(C=O)C=C1
C7H7BO3
MFCD00151823
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
(4-formylphenyl)boronic acid

Specifications

87199-17-5
100.0
96% min. (HPLC)
C7H7BO3
1g
VXWBQOJISHAKKM-UHFFFAOYSA-N
(4-formylphenyl)boronic acid
591073
97%
96.0
Authentic
Glass bottle
MFCD00151823
4-formylphenyl boronic acid, 4-formylbenzeneboronic acid, 4-boronobenzaldehyde, boronic acid, 4-formylphenyl, boronic acid, b-4-formylphenyl, p-formylphenylboronic acid, 4-dihydroxyboryl benzaldehyde, 4-dihydroxyboranyl benzaldehyde, 4-benzaldehyde boronic acid, 4-formyl-phenylboronic acid
OB(O)C1=CC=C(C=O)C=C1
149.94
149.94
4-Formylphenylboronic acid, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only