Learn More
4-Heptanone, 98%
CAS: 123-19-3 | C7H14O | 114.19 g/mol
$181.83 - $181.83
Chemical Identifiers
| CAS | 123-19-3 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.19 |
| MDL Number | MFCD00009403 |
| InChI Key | HCFAJYNVAYBARA-UHFFFAOYSA-N |
| Synonym | 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon |
| PubChem CID | 31246 |
| IUPAC Name | heptan-4-one |
| SMILES | CCCC(=O)CCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC146555000
|
Thermo Scientific Chemicals
146555000 |
500 mL | Glass bottle |
Each for $181.83
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 123-19-3 | |
| 114.19 | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| 31246 | |
| CCCC(=O)CCC |
| C7H14O | |
| MFCD00009403 | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| heptan-4-one |
Specifications
| 123-19-3 | |
| 100.0 | |
| Colorless to Yellow | |
| 145.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4060 to 1.4080 | |
| MFCD00009403 | |
| 01,699 | |
| 15,3386 | |
| Solubility in water: 4.6g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| CCCC(=O)CCC | |
| 114.19 | |
| 0.73 mPa.s (20°C) | |
| 98% | |
| 4-Heptanone, 98% |
| 97.5 | |
| -33.0°C | |
| 0.8100g/mL | |
| 48°C | |
| 97.5% min. (GC) | |
| C7H14O | |
| CH3CH2CH2COCH2CH2CH3 | |
| 500 mL | |
| 0.81 | |
| 4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon | |
| HCFAJYNVAYBARA-UHFFFAOYSA-N | |
| heptan-4-one | |
| 31246 | |
| 114.19 | |
| Liquid |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-608-9
RTECSNumber : MJ5600000
TSCA : TSCA
RUO – Research Use Only