missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3163903

 View more versions of this product

Catalog No. AAH3163903


Explore available promotions

Description

Description

As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4-Hydroxy-3-(trifluoromethyl)benzonitrile
171°C to 175°C
MFCD06797882
4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile
QXIFYONUKBXFTH-UHFFFAOYSA-N
4-hydroxy-3-(trifluoromethyl)benzonitrile
2783159
≥98%
124811-71-8
C8H4F3NO
1 g
Insoluble in water.
C1=CC(=C(C=C1C#N)C(F)(F)F)O
187.121
187.12

Chemical Identifiers

124811-71-8
187.121
QXIFYONUKBXFTH-UHFFFAOYSA-N
2783159
C1=CC(=C(C=C1C#N)C(F)(F)F)O
C8H4F3NO
MFCD06797882
4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile
4-hydroxy-3-(trifluoromethyl)benzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only