Learn More
4-Hydroxy-3-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals
$250.19 - $1059.18
Chemical Identifiers
CAS | 124811-71-8 |
---|---|
Molecular Formula | C8H4F3NO |
Molecular Weight (g/mol) | 187.121 |
MDL Number | MFCD06797882 |
InChI Key | QXIFYONUKBXFTH-UHFFFAOYSA-N |
Synonym | 4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile |
PubChem CID | 2783159 |
IUPAC Name | 4-hydroxy-3-(trifluoromethyl)benzonitrile |
SMILES | C1=CC(=C(C=C1C#N)C(F)(F)F)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH3163903
|
Thermo Scientific Chemicals
H3163903 |
1 g |
Each for $250.19
|
|
||||||
AAH3163906
|
Thermo Scientific Chemicals
H3163906 |
5 g |
Each for $1,059.18
|
|
||||||
Description
As intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAs intermediate in organic syntheses. A novel electrolyte additive for lithium nickel manganese oxide cathode of high voltage lithium ion battery. It is oxidized preferably to carbonate solvents forming a low-impedance protective film.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
124811-71-8 | |
187.121 | |
QXIFYONUKBXFTH-UHFFFAOYSA-N | |
2783159 | |
C1=CC(=C(C=C1C#N)C(F)(F)F)O |
C8H4F3NO | |
MFCD06797882 | |
4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile | |
4-hydroxy-3-(trifluoromethyl)benzonitrile |
Specifications
124811-71-8 | |
C8H4F3NO | |
1 g | |
Insoluble in water. | |
C1=CC(=C(C=C1C#N)C(F)(F)F)O | |
187.121 | |
187.12 | |
4-Hydroxy-3-(trifluoromethyl)benzonitrile |
171°C to 175°C | |
MFCD06797882 | |
4-hydroxy-3-trifluoromethyl benzonitrile, 4-hydroxy-3-trifluoromethyl-benzonitrile, pubchem4808, acmc-1c4p4, ksc514a8j, 5-cyano-2-hydroxybenzotrifluoride, benzonitrile,4-hydroxy-3-trifluoromethyl, benzonitrile, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzenecarbonitrile | |
QXIFYONUKBXFTH-UHFFFAOYSA-N | |
4-hydroxy-3-(trifluoromethyl)benzonitrile | |
2783159 | |
≥98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only