Learn More
4-Hydroxybenzoic Acid Hydrazide, 98%, Thermo Scientific Chemicals
$86.60 - $235.89
Chemical Identifiers
CAS | 5351-23-5 |
---|---|
Molecular Formula | C7H8N2O2 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00007605 |
InChI Key | ZMZGIVVRBMFZSG-UHFFFAOYSA-N |
Synonym | 4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide |
PubChem CID | 1742 |
IUPAC Name | 4-hydroxybenzohydrazide |
SMILES | C1=CC(=CC=C1C(=O)NN)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC121000250
|
Thermo Scientific Chemicals
121000250 |
25 g | Plastic bottle |
Each for $86.60
|
|
||||
AC121001000
|
Thermo Scientific Chemicals
121001000 |
100 g | Plastic bottle |
Each for $235.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5351-23-5 | |
152.15 | |
ZMZGIVVRBMFZSG-UHFFFAOYSA-N | |
1742 | |
C1=CC(=CC=C1C(=O)NN)O |
C7H8N2O2 | |
MFCD00007605 | |
4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide | |
4-hydroxybenzohydrazide |
Specifications
5351-23-5 | |
100.0 | |
Beige to White | |
98% | |
C7H8N2O2 | |
MFCD00007605 | |
10, 174 | |
Solubility in water: 26.5g/L (25°C). Other solubilities: soluble in methanol,ethanol | |
C1=CC(=CC=C1C(=O)NN)O | |
152.15 | |
152.15 | |
Fine Crystalline Powder |
97.5 | |
260.0°C to 264.0°C | |
Authentic | |
Plastic bottle | |
HOC6H4CONHNH2 | |
25 g | |
4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazide | |
ZMZGIVVRBMFZSG-UHFFFAOYSA-N | |
4-hydroxybenzohydrazide | |
1742 | |
98% | |
4-Hydroxybenzoic acid hydrazide |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye pr
GHS Signal Word: Warning
EINECSNumber : 226-326-5
RUO – Research Use Only