missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Iodoacetanilide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 393120010

 View more versions of this product

Catalog No. AC393120010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4'-Iodoacetanilide
96.0
Authentic
Glass bottle
IC6H4NHC(=O)CH3
1g
SIULLDWIXYYVCU-UHFFFAOYSA-N
N-(4-iodophenyl)acetamide
12147
97%
622-50-4
100.0
96% min. (HPLC)
C8H8INO
MFCD00016352
n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
CC(=O)NC1=CC=C(C=C1)I
261.06
261.06

Chemical Identifiers

622-50-4
261.06
SIULLDWIXYYVCU-UHFFFAOYSA-N
12147
CC(=O)NC1=CC=C(C=C1)I
C8H8INO
MFCD00016352
n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
N-(4-iodophenyl)acetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only