missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Iodoacetanilide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 622-50-4
Molecular Formula C8H8INO
Molecular Weight (g/mol) 261.06
MDL Number MFCD00016352
InChI Key SIULLDWIXYYVCU-UHFFFAOYSA-N
Synonym n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
PubChem CID 12147
IUPAC Name N-(4-iodophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC393120010
SDS View Documents
Thermo Scientific Chemicals
393120010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC393120050
SDS View Documents
Thermo Scientific Chemicals
393120050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC393120250
SDS View Documents
Thermo Scientific Chemicals
393120250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

622-50-4
261.06
SIULLDWIXYYVCU-UHFFFAOYSA-N
12147
CC(=O)NC1=CC=C(C=C1)I
C8H8INO
MFCD00016352
n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
N-(4-iodophenyl)acetamide

Specifications

622-50-4
100.0
96% min. (HPLC)
C8H8INO
MFCD00016352
n-4-iodophenyl acetamide, 4-iodoacetanilide, 4'-iodoacetanilide, p-iodoacetanilide, acetamide, n-4-iodophenyl, acetanilide, 4'-iodo, 4'-iodo-acetanilide, pubchem3293, acmc-20akb8, maybridge1_006480
CC(=O)NC1=CC=C(C=C1)I
261.06
261.06
4'-Iodoacetanilide
96.0
Authentic
Glass bottle
IC6H4NHC(=O)CH3
1g
SIULLDWIXYYVCU-UHFFFAOYSA-N
N-(4-iodophenyl)acetamide
12147
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only