missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodoaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2193618

 View more versions of this product

Catalog No. AAB2193618


Explore available promotions

Description

Description

4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).

Solubility
Slightly soluble in water.Soluble in chloroform, methanol.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications

Specifications

4-Iodoaniline
61°C to 64°C
MFCD00007848
774101
14,5027
Slightly soluble in water.Soluble in chloroform,methanol.
C1=CC(=CC=C1N)I
219.025
219.03
540-37-4
C6H6IN
50 g
Light sensitive
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
VLVCDUSVTXIWGW-UHFFFAOYSA-N
4-iodoaniline
10893
98%

Chemical Identifiers

540-37-4
219.025
VLVCDUSVTXIWGW-UHFFFAOYSA-N
10893
C1=CC(=CC=C1N)I
C6H6IN
MFCD00007848
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
4-iodoaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 208-743-4

RTECSNumber : BY3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only