Learn More
4-Iodoaniline, 98%, Thermo Scientific Chemicals
$136.22 - $509.51
Chemical Identifiers
CAS | 540-37-4 |
---|---|
Molecular Formula | C6H6IN |
Molecular Weight (g/mol) | 219.025 |
MDL Number | MFCD00007848 |
InChI Key | VLVCDUSVTXIWGW-UHFFFAOYSA-N |
Synonym | p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine |
PubChem CID | 10893 |
IUPAC Name | 4-iodoaniline |
SMILES | C1=CC(=CC=C1N)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2193618
|
Thermo Scientific Chemicals
B2193618 |
50 g |
N/A
|
|
||||||
AAB2193630
|
Thermo Scientific Chemicals
B2193630 |
250 g |
N/A
|
|
||||||
Description
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).
Solubility
Slightly soluble in water.Soluble in chloroform, methanol.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Chemical Identifiers
540-37-4 | |
219.025 | |
VLVCDUSVTXIWGW-UHFFFAOYSA-N | |
10893 | |
C1=CC(=CC=C1N)I |
C6H6IN | |
MFCD00007848 | |
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine | |
4-iodoaniline |
Specifications
540-37-4 | |
C6H6IN | |
50 g | |
Light sensitive | |
p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine | |
VLVCDUSVTXIWGW-UHFFFAOYSA-N | |
4-iodoaniline | |
10893 | |
98% |
61°C to 64°C | |
MFCD00007848 | |
774101 | |
14,5027 | |
Slightly soluble in water.Soluble in chloroform,methanol. | |
C1=CC(=CC=C1N)I | |
219.025 | |
219.03 | |
4-Iodoaniline |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 208-743-4
RTECSNumber : BY3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only