missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1195406

 View more versions of this product

Catalog No. AAA1195406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Isopropylbenzonitrile
88°C to 90°C (5 mmHg)
1.518
5 g
4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
CC(C)C1=CC=C(C=C1)C#N
145.205
145.21
13816-33-6
C10H11N
MFCD00052966
1932883
YFDJCWXBKWRDPW-UHFFFAOYSA-N
4-propan-2-ylbenzonitrile
26289
95%

Chemical Identifiers

13816-33-6
145.205
YFDJCWXBKWRDPW-UHFFFAOYSA-N
26289
CC(C)C1=CC=C(C=C1)C#N
C10H11N
MFCD00052966
4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
4-propan-2-ylbenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-492-3

RTECSNumber : DI4650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only