missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzonitrile, 95%, Thermo Scientific Chemicals

$80.95 - $1002.59

Chemical Identifiers

CAS 13816-33-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00052966
InChI Key YFDJCWXBKWRDPW-UHFFFAOYSA-N
Synonym 4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
PubChem CID 26289
IUPAC Name 4-propan-2-ylbenzonitrile
SMILES CC(C)C1=CC=C(C=C1)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1195403
View Documents
Thermo Scientific Chemicals
A1195403
1 g
Each for $80.95
 
AAA1195406
View Documents
Thermo Scientific Chemicals
A1195406
5 g
Each for $264.95
 
AAA1195414
View Documents
Thermo Scientific Chemicals
A1195414
25 g
Each for $1,002.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13816-33-6
145.205
YFDJCWXBKWRDPW-UHFFFAOYSA-N
26289
CC(C)C1=CC=C(C=C1)C#N
C10H11N
MFCD00052966
4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
4-propan-2-ylbenzonitrile

Specifications

13816-33-6
C10H11N
MFCD00052966
1932883
YFDJCWXBKWRDPW-UHFFFAOYSA-N
4-propan-2-ylbenzonitrile
26289
95%
88°C to 90°C (5 mmHg)
1.518
1 g
4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
CC(C)C1=CC=C(C=C1)C#N
145.205
145.21
4-Isopropylbenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-492-3

RTECSNumber : DI4650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only