missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylphenol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 174511000

 View more versions of this product

Catalog No. AC174511000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Isopropylphenol
99-89-8
100.0
212.0°C to 212.4°C
97.5% min. (GC)
C9H12O
MFCD00002372
06,505
Solubility in water: .. Other solubilities: soluble in 10% sodium hydroxide,soluble in alcohol and ethers
CC(C)C1=CC=C(C=C1)O
136.19
136.19
Crystalline Mass or Crystals
98%
97.5
59.0°C to 62.0°C
Authentic
Glass bottle
(CH3)2CHC6H4OH
100 g
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
YQUQWHNMBPIWGK-UHFFFAOYSA-N
4-propan-2-ylphenol
7465
98%

Chemical Identifiers

99-89-8
136.19
YQUQWHNMBPIWGK-UHFFFAOYSA-N
7465
CC(C)C1=CC=C(C=C1)O
C9H12O
MFCD00002372
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol
4-propan-2-ylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 202-798-8

RTECSNumber : SL5950000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only