Learn More
4-Isopropylphenol, 98%, Thermo Scientific Chemicals
$48.60 - $196.29
Chemical Identifiers
CAS | 99-89-8 |
---|---|
Molecular Formula | C9H12O |
Molecular Weight (g/mol) | 136.19 |
MDL Number | MFCD00002372 |
InChI Key | YQUQWHNMBPIWGK-UHFFFAOYSA-N |
Synonym | 4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol |
PubChem CID | 7465 |
IUPAC Name | 4-propan-2-ylphenol |
SMILES | CC(C)C1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC174510050
|
Thermo Scientific Chemicals
174510050 |
5 g | Glass bottle |
Each for $48.60
|
|
||||
AC174511000
|
Thermo Scientific Chemicals
174511000 |
100 g | Glass bottle |
Each for $100.59
|
|
||||
AC174512500
|
Thermo Scientific Chemicals
174512500 |
250 g | Glass bottle |
Each for $196.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-89-8 | |
136.19 | |
YQUQWHNMBPIWGK-UHFFFAOYSA-N | |
7465 | |
CC(C)C1=CC=C(C=C1)O |
C9H12O | |
MFCD00002372 | |
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol | |
4-propan-2-ylphenol |
Specifications
99-89-8 | |
100.0 | |
212.0°C to 212.4°C | |
97.5% min. (GC) | |
C9H12O | |
MFCD00002372 | |
06,505 | |
Solubility in water: .. Other solubilities: soluble in 10% sodium hydroxide,soluble in alcohol and ethers | |
CC(C)C1=CC=C(C=C1)O | |
136.19 | |
136.19 | |
Crystalline Mass or Crystals |
97.5 | |
59.0°C to 62.0°C | |
Authentic | |
Glass bottle | |
(CH3)2CHC6H4OH | |
5 g | |
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol | |
YQUQWHNMBPIWGK-UHFFFAOYSA-N | |
4-propan-2-ylphenol | |
7465 | |
98% | |
4-Isopropylphenol, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 202-798-8
RTECSNumber : SL5950000
TSCA : TSCA
RUO – Research Use Only