missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-1-butanol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5033906

 View more versions of this product

Catalog No. AAH5033906


Explore available promotions

Description

Description

4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-Methoxy-1-butanol
0.93
73°C (163°F)
C5H12O2
MFCD00040437
4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1
KOVAQMSVARJMPH-UHFFFAOYSA-N
4-methoxybutan-1-ol
8107
≥98%
111-32-0
66°C (7 mmHg)
Mild
CH3O(CH2)4OH
5 g
Slightly soluble in water.
COCCCCO
104.15
104.15

Chemical Identifiers

111-32-0
104.15
KOVAQMSVARJMPH-UHFFFAOYSA-N
8107
COCCCCO
C5H12O2
MFCD00040437
4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1
4-methoxybutan-1-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 203-858-6

RTECSNumber : EL4550000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only