Learn More
4-Methoxy-1-butanol, 98+%, Thermo Scientific Chemicals
$77.10 - $319.82
Chemical Identifiers
CAS | 111-32-0 |
---|---|
Molecular Formula | C5H12O2 |
Molecular Weight (g/mol) | 104.15 |
MDL Number | MFCD00040437 |
InChI Key | KOVAQMSVARJMPH-UHFFFAOYSA-N |
Synonym | 4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1 |
PubChem CID | 8107 |
IUPAC Name | 4-methoxybutan-1-ol |
SMILES | COCCCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5033903
|
Thermo Scientific Chemicals
H5033903 |
1 g |
Each for $77.10
|
|
|||||
AAH5033906
|
Thermo Scientific Chemicals
H5033906 |
5 g |
Each for $319.82
|
|
|||||
Description
4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
111-32-0 | |
104.15 | |
KOVAQMSVARJMPH-UHFFFAOYSA-N | |
8107 | |
COCCCCO |
C5H12O2 | |
MFCD00040437 | |
4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1 | |
4-methoxybutan-1-ol |
Specifications
111-32-0 | |
66°C (7 mmHg) | |
Mild | |
CH3O(CH2)4OH | |
1 g | |
Slightly soluble in water. | |
COCCCCO | |
104.15 | |
104.15 | |
4-Methoxy-1-butanol |
0.93 | |
73°C (163°F) | |
C5H12O2 | |
MFCD00040437 | |
4-methoxy-1-butanol, 1-butanol, 4-methoxy, dowanol bmat, butylene glycol methyl ether, dowanol bm, 4-methoxybutyl alcohol, 4-methoxybutanol, dowanal bmat, butylene glycol monomethyl ether, 4-methoxybutanol-1 | |
KOVAQMSVARJMPH-UHFFFAOYSA-N | |
4-methoxybutan-1-ol | |
8107 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 203-858-6
RTECSNumber : EL4550000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only