missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H57808MD

 View more versions of this product

Catalog No. AAH57808MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide is involved in long-chain fatty acid stimulated GLP-1 secretion and has been implicated in glucose homeostasis, adiposity, and regulation of the anti-inflammatory response. It enhances glucose-stimulated insulin secretion in MIN6 cells. It induces intracellular calcium accumulation in U2OS cells; this activity is inhibited by AH 7614.

Solubility
Soluble to 100 mM in DMSO and to 50 mM in ethanol.

Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
Specifications

Specifications

4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
C16H19NO3S
250 mg
4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide
FQUAFMNPXPXOJE-UHFFFAOYSA-N
4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
743974
97%
349085-82-1
MFCD02007969
23102756
Soluble to 100 mM in DMSO and to 50 mM in ethanol.
CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C
305.392
305.39

Chemical Identifiers

349085-82-1
305.392
FQUAFMNPXPXOJE-UHFFFAOYSA-N
743974
CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C
C16H19NO3S
MFCD02007969
4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide
4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions