missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide, 97%, Thermo Scientific™
$172.40 - $494.73
Chemical Identifiers
CAS | 349085-82-1 |
---|---|
Molecular Formula | C16H19NO3S |
Molecular Weight (g/mol) | 305.392 |
MDL Number | MFCD02007969 |
InChI Key | FQUAFMNPXPXOJE-UHFFFAOYSA-N |
Synonym | 4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide |
PubChem CID | 743974 |
IUPAC Name | 4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide |
SMILES | CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH57808MD
|
Thermo Scientific Chemicals
H57808MD |
250 mg |
Each for $172.40
|
|
|||||
AAH5780803
|
Thermo Scientific Chemicals
H5780803 |
1 g |
Each for $494.73
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide is involved in long-chain fatty acid stimulated GLP-1 secretion and has been implicated in glucose homeostasis, adiposity, and regulation of the anti-inflammatory response. It enhances glucose-stimulated insulin secretion in MIN6 cells. It induces intracellular calcium accumulation in U2OS cells; this activity is inhibited by AH 7614.
Solubility
Soluble to 100 mM in DMSO and to 50 mM in ethanol.
Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
Chemical Identifiers
349085-82-1 | |
305.392 | |
FQUAFMNPXPXOJE-UHFFFAOYSA-N | |
743974 | |
CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C |
C16H19NO3S | |
MFCD02007969 | |
4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide | |
4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide |
Specifications
349085-82-1 | |
MFCD02007969 | |
23102756 | |
Soluble to 100 mM in DMSO and to 50 mM in ethanol. | |
CC1=CC(=C(C(=C1)C)NS(=O)(=O)C2=CC=C(C=C2)OC)C | |
305.392 | |
305.39 | |
4-Methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide |
C16H19NO3S | |
250 mg | |
4-methoxy-n-2,4,6-trimethylphenyl benzenesulfonamide, cbmicro_012617, n-mesityl-4-methoxy-1-benzenesulfonamide, gsk137647a hplc, 4-methoxyphenyl sulfonyl 2,4,6-trimethylphenyl amine, 4-methoxy-n-2,4,6-trimethylphenyl benzene-1-sulfonamide | |
FQUAFMNPXPXOJE-UHFFFAOYSA-N | |
4-methoxy-N-(2,4,6-trimethylphenyl)benzenesulfonamide | |
743974 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures