missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1555936

 View more versions of this product

Catalog No. AAA1555936


Explore available promotions

Description

Description

4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

Solubility
Soluble in water, chloroform, ethyl acetate.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, acid chlorides, acid anhydrides.
Specifications

Specifications

4-Methoxybenzyl alcohol
22°C to 25°C
259°C
C8H10O2
MFCD00004653
636654
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
MSHFRERJPWKJFX-UHFFFAOYSA-N
(4-methoxyphenyl)methanol
7738
138.17
105-13-5
1.11
113°C (235°F)
1.545
500 g
14,604
Soluble in water,chloroform,ethyl acetate.
COC1=CC=C(CO)C=C1
138.17
CHEBI:86918
98%

Chemical Identifiers

105-13-5
138.17
MSHFRERJPWKJFX-UHFFFAOYSA-N
7738
(4-methoxyphenyl)methanol
C8H10O2
MFCD00004653
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
CHEBI:86918
COC1=CC=C(CO)C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c

H315-H317-H318

EINECSNumber : 203-273-6

RTECSNumber : DO8925000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only