Learn More
4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
$71.10 - $940.38
Chemical Identifiers
CAS | 105-13-5 |
---|---|
Molecular Formula | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
MDL Number | MFCD00004653 |
InChI Key | MSHFRERJPWKJFX-UHFFFAOYSA-N |
Synonym | 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol |
PubChem CID | 7738 |
ChEBI | CHEBI:86918 |
IUPAC Name | (4-methoxyphenyl)methanol |
SMILES | COC1=CC=C(CO)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1555922
|
Thermo Scientific Chemicals
A1555922 |
100 g |
Each for $71.10
|
|
|||||
AAA1555936
|
Thermo Scientific Chemicals
A1555936 |
500 g |
Each for $227.47
|
|
|||||
AAA155590E
|
Thermo Scientific Chemicals
A155590E |
2500 g |
Each for $940.38
|
|
|||||
Description
4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.
Solubility
Soluble in water, chloroform, ethyl acetate.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, acid chlorides, acid anhydrides.
Chemical Identifiers
105-13-5 | |
138.17 | |
MSHFRERJPWKJFX-UHFFFAOYSA-N | |
7738 | |
(4-methoxyphenyl)methanol |
C8H10O2 | |
MFCD00004653 | |
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
CHEBI:86918 | |
COC1=CC=C(CO)C=C1 |
Specifications
105-13-5 | |
1.11 | |
113°C (235°F) | |
1.545 | |
100 g | |
14,604 | |
Soluble in water,chloroform,ethyl acetate. | |
COC1=CC=C(CO)C=C1 | |
138.17 | |
CHEBI:86918 | |
98% |
22°C to 25°C | |
259°C | |
C8H10O2 | |
MFCD00004653 | |
636654 | |
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
MSHFRERJPWKJFX-UHFFFAOYSA-N | |
(4-methoxyphenyl)methanol | |
7738 | |
138.17 | |
4-Methoxybenzyl alcohol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
H315-H317-H318
EINECSNumber : 203-273-6
RTECSNumber : DO8925000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only