missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2H-1,4-benzoxazin-3(4H)-one, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2719614

 View more versions of this product

Catalog No. AAH2719614


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methyl-2H-1,4-benzoxazin-3(4H)-one
58°C to 60°C
MFCD00601365
4-methyl-2h-1,4-benzoxazin-3 4h-one, 4-methyl-2h-1,4-benzoxazin-3-one, 4-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,4-methyl, 4-methyl-4h-benzo 1,4 oxazin-3-one, 4-methyl-2h-benzo e 1,4-oxazaperhydroin-3-one, acmc-20ame8, 4-methyl-2h-1,4-benzoxazin-3-4h-one, 2h-1,4-benzoxazin-3 4h-one, 4-methyl
CN1C(=O)COC2=CC=CC=C21
163.176
163.18
21744-84-3
C9H9NO2
25 g
DBJMEBUKQVZWMD-UHFFFAOYSA-N
4-methyl-1,4-benzoxazin-3-one
89032
98%

Chemical Identifiers

21744-84-3
163.176
DBJMEBUKQVZWMD-UHFFFAOYSA-N
4-methyl-1,4-benzoxazin-3-one
C9H9NO2
MFCD00601365
89032
CN1C(=O)COC2=CC=CC=C21
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only