missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methyl-2H-1,4-benzoxazin-3(4H)-one, 98%, Thermo Scientific Chemicals

$54.62 - $182.44

Chemical Identifiers

CAS 21744-84-3
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00601365
InChI Key DBJMEBUKQVZWMD-UHFFFAOYSA-N
PubChem CID 89032
IUPAC Name 4-methyl-1,4-benzoxazin-3-one
SMILES CN1C(=O)COC2=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2719606
View Documents
Thermo Scientific Chemicals
H2719606
5 g
Each for $54.62
 
AAH2719614
View Documents
Thermo Scientific Chemicals
H2719614
25 g
Each for $182.44
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21744-84-3
163.176
DBJMEBUKQVZWMD-UHFFFAOYSA-N
4-methyl-1,4-benzoxazin-3-one
C9H9NO2
MFCD00601365
89032
CN1C(=O)COC2=CC=CC=C21

Specifications

21744-84-3
C9H9NO2
5 g
DBJMEBUKQVZWMD-UHFFFAOYSA-N
4-methyl-1,4-benzoxazin-3-one
89032
98%
58°C to 60°C
MFCD00601365
4-methyl-2h-1,4-benzoxazin-3 4h-one, 4-methyl-2h-1,4-benzoxazin-3-one, 4-methyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,4-methyl, 4-methyl-4h-benzo 1,4 oxazin-3-one, 4-methyl-2h-benzo e 1,4-oxazaperhydroin-3-one, acmc-20ame8, 4-methyl-2h-1,4-benzoxazin-3-4h-one, 2h-1,4-benzoxazin-3 4h-one, 4-methyl
CN1C(=O)COC2=CC=CC=C21
163.176
163.18
4-Methyl-2H-1,4-benzoxazin-3(4H)-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only