missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Morpholin-4-yl-phenyl)-methanol, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC17409EA
Specifications
(4-Morpholin-4-yl-phenyl)-methanol | |
95% | |
C11H15NO2 | |
10g | |
KUAHZNZWSIDSTH-UHFFFAOYSA-N | |
(4-morpholin-4-ylphenyl)methanol | |
2776456 | |
95% |
280556-71-0 | |
Amber Glass Bottle | |
MFCD01057413 | |
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol | |
OCC1=CC=C(C=C1)N1CCOCC1 | |
193.25 | |
193.24 |
Chemical Identifiers
280556-71-0 | |
193.25 | |
KUAHZNZWSIDSTH-UHFFFAOYSA-N | |
2776456 | |
OCC1=CC=C(C=C1)N1CCOCC1 |
C11H15NO2 | |
MFCD01057413 | |
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol | |
(4-morpholin-4-ylphenyl)methanol |
Safety and Handling
GHS Signal Word: Warning