missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Morpholin-4-yl-phenyl)-methanol, 95+%, Thermo Scientific™
Chemical Identifiers
CAS | 280556-71-0 |
---|---|
Molecular Formula | C11H15NO2 |
Molecular Weight (g/mol) | 193.25 |
MDL Number | MFCD01057413 |
InChI Key | KUAHZNZWSIDSTH-UHFFFAOYSA-N |
Synonym | 4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol |
PubChem CID | 2776456 |
IUPAC Name | (4-morpholin-4-ylphenyl)methanol |
SMILES | OCC1=CC=C(C=C1)N1CCOCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC17409CB
|
Thermo Scientific Chemicals
CC17409CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC17409DA
|
Thermo Scientific Chemicals
CC17409DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC17409EA
|
Thermo Scientific Chemicals
CC17409EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
280556-71-0 | |
193.25 | |
KUAHZNZWSIDSTH-UHFFFAOYSA-N | |
2776456 | |
OCC1=CC=C(C=C1)N1CCOCC1 |
C11H15NO2 | |
MFCD01057413 | |
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol | |
(4-morpholin-4-ylphenyl)methanol |
Specifications
280556-71-0 | |
Amber Glass Bottle | |
MFCD01057413 | |
4-morpholin-4-yl-phenyl methanol, 4-morpholinobenzyl alcohol, 4-morpholin-4-yl phenyl methanol, 4-morpholinophenyl methanol, 4-morpholin-4-yl-phenyl-methanol, 4-morpholin-4-ylphenyl methanol, 4-morpholin-4-yl benzyl alcohol, 4-morpholin-4-ylphenyl methan-1-ol, 4-morpholinobenzylalcohol, 4-4-morpholinyl benzenemethanol | |
OCC1=CC=C(C=C1)N1CCOCC1 | |
193.25 | |
193.24 | |
(4-Morpholin-4-yl-phenyl)-methanol |
95% | |
C11H15NO2 | |
250mg | |
KUAHZNZWSIDSTH-UHFFFAOYSA-N | |
[4-(morpholin-4-yl)phenyl]methanol | |
2776456 | |
95% |
Safety and Handling
GHS Signal Word: Warning