missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-n-Butoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1125803

 View more versions of this product

Catalog No. AAL1125803


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-n-Butoxybenzenesulfonyl chloride
130°C to 131°C (0.6 mmHg)
1.542
1 g
1109264
4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54
CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl
248.721
248.73
1138-56-3
C10H13ClO3S
MFCD00052344
UN3265
Moisture sensitive
HGKWMUBXVMFXNC-UHFFFAOYSA-N
4-butoxybenzenesulfonyl chloride
517990
97%

Chemical Identifiers

1138-56-3
248.721
HGKWMUBXVMFXNC-UHFFFAOYSA-N
517990
CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl
C10H13ClO3S
MFCD00052344
4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54
4-butoxybenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only