Learn More
4-n-Butoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
$50.78 - $430.19
Chemical Identifiers
CAS | 1138-56-3 |
---|---|
Molecular Formula | C10H13ClO3S |
Molecular Weight (g/mol) | 248.721 |
MDL Number | MFCD00052344 |
InChI Key | HGKWMUBXVMFXNC-UHFFFAOYSA-N |
Synonym | 4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54 |
PubChem CID | 517990 |
IUPAC Name | 4-butoxybenzenesulfonyl chloride |
SMILES | CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL11258MD
|
Thermo Scientific Chemicals
L11258MD |
250 mg |
Each for $50.78
|
|
|||||
AAL1125803
|
Thermo Scientific Chemicals
L1125803 |
1 g |
Each for $131.63
|
|
|||||
AAL1125806
|
Thermo Scientific Chemicals
L1125806 |
5 g |
Each for $430.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1138-56-3 | |
248.721 | |
HGKWMUBXVMFXNC-UHFFFAOYSA-N | |
517990 | |
CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl |
C10H13ClO3S | |
MFCD00052344 | |
4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54 | |
4-butoxybenzenesulfonyl chloride |
Specifications
1138-56-3 | |
C10H13ClO3S | |
MFCD00052344 | |
UN3265 | |
Moisture sensitive | |
HGKWMUBXVMFXNC-UHFFFAOYSA-N | |
4-butoxybenzenesulfonyl chloride | |
517990 | |
97% |
130°C to 131°C (0.6 mmHg) | |
1.542 | |
250 mg | |
1109264 | |
4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54 | |
CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl | |
248.721 | |
248.73 | |
4-n-Butoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only