Learn More
4-n-Butoxyphenyl isocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1287703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-n-Butoxyphenyl isocyanate | |
1.055 | |
>110°C (230°F) | |
1.522 | |
1 g | |
2107851 | |
4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate | |
CCCCOC1=CC=C(C=C1)N=C=O | |
191.23 | |
191.23 |
28439-86-3 | |
149°C to 150°C (3 mmHg) | |
C11H13NO2 | |
MFCD00037042 | |
UN2206 | |
Moisture sensitive | |
ASFIDVOMDQBCNP-UHFFFAOYSA-N | |
1-butoxy-4-isocyanatobenzene | |
2769629 | |
98% |
Chemical Identifiers
28439-86-3 | |
191.23 | |
ASFIDVOMDQBCNP-UHFFFAOYSA-N | |
2769629 | |
CCCCOC1=CC=C(C=C1)N=C=O |
C11H13NO2 | |
MFCD00037042 | |
4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate | |
1-butoxy-4-isocyanatobenzene |
Safety and Handling
GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302+H332-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only