missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-n-Butoxyphenyl isocyanate, 98%, Thermo Scientific Chemicals

$65.37 - $239.99

Chemical Identifiers

CAS 28439-86-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00037042
InChI Key ASFIDVOMDQBCNP-UHFFFAOYSA-N
Synonym 4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
PubChem CID 2769629
IUPAC Name 1-butoxy-4-isocyanatobenzene
SMILES CCCCOC1=CC=C(C=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1287703
View Documents
Thermo Scientific Chemicals
L1287703
1 g
Each for $65.37
 
AAL1287706
View Documents
Thermo Scientific Chemicals
L1287706
5 g
Each for $239.99
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28439-86-3
191.23
ASFIDVOMDQBCNP-UHFFFAOYSA-N
2769629
CCCCOC1=CC=C(C=C1)N=C=O
C11H13NO2
MFCD00037042
4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
1-butoxy-4-isocyanatobenzene

Specifications

28439-86-3
149°C to 150°C (3 mmHg)
C11H13NO2
MFCD00037042
UN2206
Moisture sensitive
ASFIDVOMDQBCNP-UHFFFAOYSA-N
1-butoxy-4-isocyanatobenzene
2769629
98%
1.055
>110°C (230°F)
1.522
1 g
2107851
4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
CCCCOC1=CC=C(C=C1)N=C=O
191.23
191.23
4-n-Butoxyphenyl isocyanate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H302-H332-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302+H332-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only