missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-n-Pentylphenylacetylene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6091309

 View more versions of this product

Catalog No. AAH6091309


Explore available promotions

Description

Description

4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-n-Pentylphenylacetylene
0.885
103°C (217°F)
1.523
10 g
APGNXGIUUTWIRE-UHFFFAOYSA-N
1-ethynyl-4-pentylbenzene
2775131
97%
79887-10-8
172°C
C13H16
MFCD00173884
4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
CCCCCC1=CC=C(C=C1)C#C
172.27
172.27

Chemical Identifiers

79887-10-8
172.27
APGNXGIUUTWIRE-UHFFFAOYSA-N
2775131
CCCCCC1=CC=C(C=C1)C#C
C13H16
MFCD00173884
4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
1-ethynyl-4-pentylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only