missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-n-Pentylphenylacetylene, 97%, Thermo Scientific Chemicals
$58.74 - $218.12
Chemical Identifiers
CAS | 79887-10-8 |
---|---|
Molecular Formula | C13H16 |
Molecular Weight (g/mol) | 172.27 |
MDL Number | MFCD00173884 |
InChI Key | APGNXGIUUTWIRE-UHFFFAOYSA-N |
Synonym | 4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene |
PubChem CID | 2775131 |
IUPAC Name | 1-ethynyl-4-pentylbenzene |
SMILES | CCCCCC1=CC=C(C=C1)C#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6091304
|
Thermo Scientific Chemicals
H6091304 |
2 g |
Each for $58.74
|
|
|||||
AAH6091309
|
Thermo Scientific Chemicals
H6091309 |
10 g |
Each for $218.12
|
|
|||||
Description
4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
Chemical Identifiers
79887-10-8 | |
172.27 | |
APGNXGIUUTWIRE-UHFFFAOYSA-N | |
2775131 | |
CCCCCC1=CC=C(C=C1)C#C |
C13H16 | |
MFCD00173884 | |
4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene | |
1-ethynyl-4-pentylbenzene |
Specifications
79887-10-8 | |
172°C | |
C13H16 | |
MFCD00173884 | |
4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene | |
CCCCCC1=CC=C(C=C1)C#C | |
172.27 | |
172.27 | |
4-n-Pentylphenylacetylene |
0.885 | |
103°C (217°F) | |
1.523 | |
2 g | |
APGNXGIUUTWIRE-UHFFFAOYSA-N | |
1-ethynyl-4-pentylbenzene | |
2775131 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only