missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophenyl benzoate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1602414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Nitrophenyl benzoate | |
142°C to 144°C | |
MFCD00135494 | |
2054709 | |
GMKZBFFLCONHDE-UHFFFAOYSA-N | |
(4-nitrophenyl) benzoate | |
70396 | |
97% |
959-22-8 | |
C13H9NO4 | |
25 g | |
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester | |
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-] | |
243.218 | |
243.22 |
Chemical Identifiers
959-22-8 | |
243.218 | |
GMKZBFFLCONHDE-UHFFFAOYSA-N | |
70396 | |
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-] |
C13H9NO4 | |
MFCD00135494 | |
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester | |
(4-nitrophenyl) benzoate |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only