Learn More
4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 296700050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Nitrophenyl isocyanate | |
97.5 | |
56.0°C to 62.0°C | |
>110°C | |
97.5% min. (GC) | |
C7H4N2O3 | |
5 g | |
Solubility in water: .. Other solubilities: soluble in benzene and toluene | |
C1=CC(=CC=C1N=C=O)[N+](=O)[O-] | |
164.12 | |
164.12 | |
Crystalline Solid |
100-28-7 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
O2NC6H4NCO | |
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate | |
GFNKTLQTQSALEJ-UHFFFAOYSA-N | |
1-isocyanato-4-nitrobenzene | |
66012 | |
98% |
Chemical Identifiers
100-28-7 | |
164.12 | |
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate | |
1-isocyanato-4-nitrobenzene |
C7H4N2O3 | |
GFNKTLQTQSALEJ-UHFFFAOYSA-N | |
66012 | |
C1=CC(=CC=C1N=C=O)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 202-836-3
RUO – Research Use Only