Learn More
4-Nitrophenyl isocyanate, 98%, Thermo Scientific Chemicals
$60.71 - $205.62
Chemical Identifiers
CAS | 100-28-7 |
---|---|
Molecular Formula | C7H4N2O3 |
Molecular Weight (g/mol) | 164.12 |
InChI Key | GFNKTLQTQSALEJ-UHFFFAOYSA-N |
Synonym | 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate |
PubChem CID | 66012 |
IUPAC Name | 1-isocyanato-4-nitrobenzene |
SMILES | C1=CC(=CC=C1N=C=O)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC296700050
|
Thermo Scientific Chemicals
296700050 |
5 g | Glass bottle |
Each for $60.71
|
|
||||
AC296700250
|
Thermo Scientific Chemicals
296700250 |
25 g | Glass bottle |
Each for $205.62
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-28-7 | |
164.12 | |
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate | |
1-isocyanato-4-nitrobenzene |
C7H4N2O3 | |
GFNKTLQTQSALEJ-UHFFFAOYSA-N | |
66012 | |
C1=CC(=CC=C1N=C=O)[N+](=O)[O-] |
Specifications
100-28-7 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
O2NC6H4NCO | |
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate | |
GFNKTLQTQSALEJ-UHFFFAOYSA-N | |
1-isocyanato-4-nitrobenzene | |
66012 | |
98% | |
4-Nitrophenyl isocyanate |
97.5 | |
56.0°C to 62.0°C | |
>110°C | |
97.5% min. (GC) | |
C7H4N2O3 | |
5 g | |
Solubility in water: .. Other solubilities: soluble in benzene and toluene | |
C1=CC(=CC=C1N=C=O)[N+](=O)[O-] | |
164.12 | |
164.12 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 202-836-3
RUO – Research Use Only