missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6646503
Description
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.
Solubility
Soluble in water
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
4-Nitrophenylphosphorylcholine | |
233°C to 235°C | |
C11H17N2O6P | |
1 g | |
Light and moisture sensitive | |
Soluble in water | |
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O | |
304.24 | |
304.24 |
21064-69-7 | |
Yellow | |
MFCD00077834 | |
1895422 | |
p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt | |
NAIXASFEPQPICN-UHFFFAOYSA-N | |
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate | |
4307994 | |
Solid |
Chemical Identifiers
21064-69-7 | |
304.24 | |
NAIXASFEPQPICN-UHFFFAOYSA-N | |
4307994 | |
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O |
C11H17N2O6P | |
MFCD00077834 | |
p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt | |
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only