missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenylphosphorylcholine, Thermo Scientific Chemicals

$106.41 - $805.15

Chemical Identifiers

CAS 21064-69-7
Molecular Formula C11H17N2O6P
Molecular Weight (g/mol) 304.24
MDL Number MFCD00077834
InChI Key NAIXASFEPQPICN-UHFFFAOYSA-N
Synonym p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt
PubChem CID 4307994
IUPAC Name (4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66465MC
View Documents
Thermo Scientific Chemicals
J66465MC
100 mg
Each for $106.41
 
AAJ66465ME
View Documents
Thermo Scientific Chemicals
J66465ME
500 mg
Each for $496.01
 
AAJ6646503
View Documents
Thermo Scientific Chemicals
J6646503
1 g
Each for $805.15
 
Description

Description

O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of Phospholipase C.

Solubility
Soluble in water

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

21064-69-7
304.24
NAIXASFEPQPICN-UHFFFAOYSA-N
4307994
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
C11H17N2O6P
MFCD00077834
p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate

Specifications

21064-69-7
Yellow
MFCD00077834
1895422
p-nitrophenylphosphorylcholine, o-4-nitrophenylphosphoryl choline, 4-nitrophenylphosphorylcholine, 4-nitrophenyl 2-trimethylammonio ethyl phosphate, 4-nitrophenylphorylcholine, o-4-nitrophenyl-phosphoryl choline, 4-nitrophenyl 2-trimethylazaniumyl ethyl phosphate, 2-hydroxy 4-nitrophenoxy phosphinyl oxy-n,n,n-trimethyl-ethanaminium inner salt
NAIXASFEPQPICN-UHFFFAOYSA-N
(4-nitrophenyl) 2-(trimethylazaniumyl)ethyl phosphate
4307994
Solid
233°C to 235°C
C11H17N2O6P
100 mg
Light and moisture sensitive
Soluble in water
C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
304.24
304.24
4-Nitrophenylphosphorylcholine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only