missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophthalic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 416121000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Nitrophthalic acid | |
Authentic | |
Glass bottle | |
100g | |
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O | |
69121 | |
99% |
610-27-5 | |
98.5% min. (HPLC) | |
C8H5NO6 | |
09,828 | |
SLBQXWXKPNIVSQ-UHFFFAOYSA-N | |
211.13 | |
211.13 |
Chemical Identifiers
610-27-5 | |
211.13 | |
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O |
C8H5NO6 | |
SLBQXWXKPNIVSQ-UHFFFAOYSA-N | |
69121 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 210-215-3
RUO â Research Use Only