missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophthalic acid, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 610-27-5 |
---|---|
Molecular Formula | C8H5NO6 |
Molecular Weight (g/mol) | 211.13 |
InChI Key | SLBQXWXKPNIVSQ-UHFFFAOYSA-N |
Synonym | 4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8 |
PubChem CID | 69121 |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC416120250
|
Thermo Scientific Chemicals
416120250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC416121000
|
Thermo Scientific Chemicals
416121000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
610-27-5 | |
211.13 | |
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O |
C8H5NO6 | |
SLBQXWXKPNIVSQ-UHFFFAOYSA-N | |
69121 |
Specifications
610-27-5 | |
98.5% min. (HPLC) | |
C8H5NO6 | |
09,828 | |
SLBQXWXKPNIVSQ-UHFFFAOYSA-N | |
211.13 | |
211.13 | |
4-Nitrophthalic acid |
Authentic | |
Glass bottle | |
25g | |
4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8 | |
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O | |
69121 | |
99% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 210-215-3
RUO â Research Use Only