missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Penten-1-ol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1117822

 View more versions of this product

Catalog No. AAA1117822


Explore available promotions

Description

Description

4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.

Solubility
Solubility in water: 57 g/L (25°C)

Notes
Protect against electrostatic charges. Store away from oxidizing agents. Use only in a chemical fume hood. Store at 2-8°C.
Specifications

Specifications

4-Penten-1-ol
0.84
43°C (109°F)
1.43
100 g
1560163
LQAVWYMTUMSFBE-UHFFFAOYSA-N
pent-4-en-1-ol
13181
≥98%
821-09-0
134°C to 137°C
C5H10O
MFCD00002975
UN1987
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
C=CCCCO
86.134
86.13
Liquid

Chemical Identifiers

821-09-0
86.134
LQAVWYMTUMSFBE-UHFFFAOYSA-N
13181
C=CCCCO
C5H10O
MFCD00002975
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene
pent-4-en-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 212-473-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only