Learn More
4-Penten-1-ol, 98+%
CAS: 821-09-0 | C5H10O | 86.134 g/mol
$121.29 - $1232.18
Chemical Identifiers
| CAS | 821-09-0 |
|---|---|
| Molecular Formula | C5H10O |
| Molecular Weight (g/mol) | 86.134 |
| MDL Number | MFCD00002975 |
| InChI Key | LQAVWYMTUMSFBE-UHFFFAOYSA-N |
| Synonym | 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene |
| PubChem CID | 13181 |
| IUPAC Name | pent-4-en-1-ol |
| SMILES | C=CCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1117806
|
Thermo Scientific Chemicals
A1117806 |
5 g |
Each for $121.29
|
|
|||||
|
AAA1117814
|
Thermo Scientific Chemicals
A1117814 |
25 g |
Each for $400.73
|
|
|||||
|
AAA1117822
|
Thermo Scientific Chemicals
A1117822 |
100 g |
Each for $1,232.18
|
|
|||||
Description
4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Penten-1-ol is used as a reagent used in carbohydrate chemistry for n-Pentenyl Glycoside methodology in the rapid assembly of homoglycans exemplified with the nonasaccharide component of a high-mannose glycoprotein. It can be used in agrochemical, pharmaceutical and dyestuff field etc. 4-Penten-1-ol is used to study the epoxidation of olefins with oxo-diperoxo tungstate(VI) complex as catalyst and bicarbonate as co-catalyst.
Solubility
Solubility in water: 57 g/L (25°C)
Notes
Protect against electrostatic charges. Store away from oxidizing agents. Use only in a chemical fume hood. Store at 2-8°C.
Chemical Identifiers
| 821-09-0 | |
| 86.134 | |
| LQAVWYMTUMSFBE-UHFFFAOYSA-N | |
| 13181 | |
| C=CCCCO |
| C5H10O | |
| MFCD00002975 | |
| 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene | |
| pent-4-en-1-ol |
Specifications
| 821-09-0 | |
| 134°C to 137°C | |
| C5H10O | |
| MFCD00002975 | |
| UN1987 | |
| 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene | |
| C=CCCCO | |
| 86.134 | |
| 86.13 | |
| Liquid |
| 0.84 | |
| 43°C (109°F) | |
| 1.43 | |
| 5 g | |
| 1560163 | |
| LQAVWYMTUMSFBE-UHFFFAOYSA-N | |
| pent-4-en-1-ol | |
| 13181 | |
| ≥98% | |
| 4-Penten-1-ol |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 212-473-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only